2-Butynoic acid methyl ester 98%

Supplier: Ambeed
Danger

Synonyms: Methyl 2-butynoate, Methyl but-2-ynoate

A760588-1G A760588-25G A760588-5G
76807-238EA 10.61 USD
76807-238 76807-240 76807-242
2-Butynoic acid methyl ester 98%
2-Butynoic acid methyl ester

Precaución: for Research Use Only

Formula: C₅H₆O₂
MW: 98.1 g/mol
MDL Number: MFCD00009273
CAS Number: 23326-27-4
UN: 3272
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR