Fmoc-Piz·HCl 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(9H-Fluoren-9-yl)methyl piperazine-1-carboxylate hydrochloride
A760276-10G
A760276-1G
A760276-25G
A760276-5G
76807-226EA
125.19
USD
76807-226
76807-228
76807-230
76807-232
Fmoc-Piz·HCl 98%
Fmoc-Piz·HCl
Precaución: for Research Use Only
Formula:
C₁₉H₂₀N₂O₂·HCl MW: 344.80 g/mol |
MDL Number:
MFCD00673790 CAS Number: 215190-22-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...