p-Xylene-bis(N-pyridinium bromide) (DPX) 98%

Supplier: Ambeed
Warning

Synonyms: 1,1'-(p-Xylylene)bis(1-pyridinium) dibromide, 1,1'-(1,4-Phenylenebis(methylene))bis(pyridin-1-ium) bromide, DPX, 1,1'-(p-Xylylene)bis(1-pyridinium bromide), 1,1'-[1,4-Phenylenebis(methylene)]bis(1-pyridinium) dibromide

A759727-100MG A759727-1G
76807-222EA 33 USD
76807-222 76807-224
p-Xylene-bis(N-pyridinium bromide) (DPX) 98%
p-Xylene-bis(N-pyridinium bromide) (DPX)

Precaución: for Research Use Only

Formula: C₁₈H₁₈Br₂N₂
MDL Number: MFCD00043749
CAS Number: 14208-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR