2-(2-Butoxyethoxy)ethyl acetate 97%

Supplier: Ambeed
Warning

Synonyms: Diethyleneglycolmonobutyletheracetate, Diethylene glycol monobutyl ether acetate

A757722-1000G A757722-100G A757722-25G A757722-500G
76807-208EA 23 USD
76807-208 76807-210 76807-212 76807-214
2-(2-Butoxyethoxy)ethyl acetate 97%
2-(2-Butoxyethoxy)ethyl acetate

Precaución: for Research Use Only

Formula: C₁₀H₂₀O₄
MDL Number: MFCD00009458
CAS Number: 124-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR