2,3-Quinoline dicarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2,3-Quinolinedicarboxylic acid

A757126-100G A757126-1G A757126-25G A757126-5G
76807-200EA 71.09 USD
76807-200 76807-202 76807-204 76807-206
2,3-Quinoline dicarboxylic acid 97%
2,3-Quinoline dicarboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₇NO₄
MW: 217.18 g/mol
MDL Number: MFCD00071756
CAS Number: 643-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR