Isethionic acid ammonium salt (ammonium 2-hydroxyethanesulfonate) 99%

Supplier: Ambeed
Warning

Synonyms: Isethionic acid ammonium salt, Ammonium 2-hydroxyethanesulphonate

A754812-1000G A754812-100G A754812-25G A754812-500G
76807-184EA 246.13 USD
76807-184 76807-186 76807-188 76807-190
Isethionic acid ammonium salt (ammonium 2-hydroxyethanesulfonate) 99%
Isethionic acid ammonium salt (ammonium 2-hydroxyethanesulphonate)

Precaución: for Research Use Only

Formula: C₂H₉NO₄S
MW: 143.16 g/mol
MDL Number: MFCD00012847
CAS Number: 57267-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR