2-Hydroxyflutamide 98%

Supplier: Ambeed
Warning

A754229-100MG A754229-250MG A754229-25MG A754229-5G A754229-1G A754229-1MG A754229-10G
76807-176EA 19 USD
76807-176 76807-178 76807-180 76807-182 77573-424 77670-030 77880-531
2-Hydroxyflutamide 98%
2-Hydroxyflutamide

Precaución: for Research Use Only

Formula: C₁₁H₁₁F₃N₂O₄
MDL Number: MFCD00563126
CAS Number: 52806-53-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR