Heptyl acetate 99%

Supplier: Ambeed
Warning

Synonyms: Acetic acid heptyl ester

A753203-100G A753203-25G A753203-500G
76807-164EA 18.04 USD
76807-164 76807-166 76807-168
Heptyl acetate 99%
Heptyl acetate

Precaución: for Research Use Only

Formula: C₉H₁₈O₂
MW: 158.24 g/mol
MDL Number: MFCD00027311
CAS Number: 112-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR