Fmoc-asp-ofm 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Asp-Ofm

A752487-1G A752487-250MG A752487-5G
76807-156EA 39.26 USD
76807-156 76807-158 76807-160
Fmoc-asp-ofm 98%
Fmoc-asp-ofm

Precaución: for Research Use Only

Formula: C₃₃H₂₇NO₆
MW: 533.58 g/mol
MDL Number: MFCD00237653
CAS Number: 187671-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR