Gold(III) oxide 99.99% Au 88.6% min

Supplier: Ambeed
Warning

A749554-100MG A749554-1G A749554-250MG
76807-140EA 51 USD
76807-140 76807-142 76807-144
Gold(III) oxide 99.99% Au 88.6% min
Gold(III) oxide

Precaución: for Research Use Only

Formula: Au₂O₃
MW: 441.93
Melting Pt: 150
Density: 3.6
MDL Number: MFCD00014173
CAS Number: 1303-58-8
Merck Index: 12,04545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR