Ethyl-O-anisate 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxybenzoic acid ethyl ester, Ethyl 2-methoxybenzoate, o-Anisic acid ethyl ester, 2-Anisic acid ethyl ester

A742448-100G A742448-25G A742448-5G
76807-112EA 130.5 USD
76807-112 76807-114 76807-116
Ethyl-O-anisate 98%
Ethyl-O-anisate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
MDL Number: MFCD00015148
CAS Number: 7335-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR