meso-1,2-Diphenylethylenediamine 97%

Supplier: Ambeed
Warning

Synonyms: meso-1,2-Diphenylethane-1,2-diamine

A741911-1G A741911-25G A741911-5G
76807-106EA 36.08 USD
76807-106 76807-108 76807-110
meso-1,2-Diphenylethylenediamine 97%
meso-1,2-Diphenylethylenediamine

Precaución: for Research Use Only

Formula: C₁₄H₁₆N₂
MW: 212.30 g/mol
MDL Number: MFCD00274328
CAS Number: 951-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR