1,4-Dihydrazinylphthalazine sulfate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Dihydralazine sulphate
A738601-100MG
A738601-1MG
A738601-25MG
A738601-50MG
A738601-5MG
A738601-10MG
A738601-250MG
76807-080EA
270
USD
76807-080
76807-082
76807-084
76807-086
76807-088
77602-896
77880-509
1,4-Dihydrazinylphthalazine sulfate 98%
1,4-Dihydrazinylphthalazine sulfate
Precaución: for Research Use Only
Formula:
C₈H₁₂N₆O₄S |
MDL Number:
MFCD00035271 CAS Number: 7327-87-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...