Methyl-1-benzyl-5-oxopyrrolidine-3-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 1-Benzyl-5-oxo-3-pyrrolidinecarboxylate

A737947-100G A737947-1G A737947-25G A737947-5G
76807-064EA 98 USD
76807-064 76807-066 76807-068 76807-070
Methyl-1-benzyl-5-oxopyrrolidine-3-carboxylate 98%
Methyl-1-benzyl-5-oxopyrrolidine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₃
MW: 233.267 g/mol
MDL Number: MFCD00003196
CAS Number: 51535-00-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR