1-Chloro-4-nitronaphthalene 98%

Supplier: Ambeed
Warning

A737060-1G A737060-250MG A737060-5G
76807-052EA 35 USD
76807-052 76807-054 76807-056
1-Chloro-4-nitronaphthalene 98%
1-Chloro-4-nitronaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₆ClNO₂
MDL Number: MFCD00465088
CAS Number: 605-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR