Manganese(ii)hexafluoroacetylacetonate 95% + (contains < 5% H₂O)

Supplier: Ambeed
Warning

A734023-1G A734023-250MG A734023-5G
76807-026EA 55.17 USD
76807-026 76807-028 76807-030
Manganese(ii)hexafluoroacetylacetonate 95% + (contains < 5% H₂O)
Manganese(ii)hexafluoroacetylacetonate

Precaución: for Research Use Only

Formula: C₁₀H₂F₁₂MnO₄
MW: 469.04 g/mol
MDL Number: MFCD09763681
CAS Number: 19648-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR