Guanosine 5'-triphosphate disodium salt 89%
Supplier: Ambeed
|
Warning
|
Synonyms:
((2R,3S,4R,5R)-5-(2-Amino-6-oxo-3,6-dihydro-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt, Guanosine-5'-triphosphate disodium salt, GTP disodium salt, Guanosine-5-triphosphoric aicd disodium salt
A731559-1G
A731559-250MG
A731559-25G
76806-992EA
41
USD
76806-992
76806-994
76806-996
Guanosine 5'-triphosphate disodium salt 89%
Guanosine 5'-triphosphate disodium salt
Precaución: for Research Use Only
Formula:
C₁₀H₁₄N₅Na₂O₁₄P₃ MW: 567.147 g/mol |
MDL Number:
MFCD03410297 CAS Number: 56001-37-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...