Diclazuril 98%

Supplier: Ambeed
Danger

Synonyms: 2-(4-Chlorophenyl)-2-(2,6-dichloro-4-(3,5-dioxo-4,5-dihydro-1,2,4-triazin-2(3H)-yl)phenyl)acetonitrile, 2,6-Dichloro-alpha-(4-chlorophenyl)-4-[4,5-dihydro-3,5-dioxo-1,2,4-triazin-2(3H)-yl]benzeneacetonitrile

A730578-100MG A730578-1G A730578-1MG A730578-250MG A730578-25G A730578-5G
76806-978EA 8 USD
76806-978 76806-980 76806-982 76806-984 76806-986 76806-988
Diclazuril 98%
Diclazuril

Precaución: for Research Use Only

Formula: C₁₇H₉Cl₃N₄O₂
MW: 407.64 g/mol
MDL Number: MFCD00867203
CAS Number: 101831-37-2
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR