(±)-Camphor-10-sulfonic acid 99%

Supplier: Ambeed
Danger

Synonyms: DL-Camphor-10-sulphonic acid, (±)-2-Oxobornane-10-sulphonic acid, (7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulphonic acid

A730355-100G A730355-25G
76806-976EA 8.49 USD
76806-976 77308-224
(±)-Camphor-10-sulfonic acid 99%
(±)-Camphor-10-sulphonic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₄S
MW: 232.3 g/mol
MDL Number: MFCD00074827
CAS Number: 5872-08-2
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR