Glycine 7-amido-4-methylcoumarin hydrobromide salt 98%

Supplier: Ambeed
Warning

A728480-100MG A728480-1G A728480-250MG
76806-968EA 110 USD
76806-968 76806-970 77880-492
Glycine 7-amido-4-methylcoumarin hydrobromide salt 98%
Glycine 7-amido-4-methylcoumarin hydrobromide salt

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O₃·BrH
MDL Number: MFCD00058452
CAS Number: 113728-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR