N-Acetyl-DL-4-fluorophenylalanine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Acetamido-3-(4-fluorophenyl)propanoic acid, N-Ac-DL-Phe(4-F)-OH

A727686-1G A727686-5G
76806-964EA 19 USD
76806-964 76806-966
N-Acetyl-DL-4-fluorophenylalanine 98%
N-Acetyl-DL-4-fluorophenylalanine

Precaución: for Research Use Only

Formula: C₁₁H₁₂FNO₃
MW: 225.22 g/mol
MDL Number: MFCD00017983
CAS Number: 17481-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR