Geranyl formate 85%

Supplier: Ambeed
Warning

A726071-100G A726071-25G A726071-500G
76806-954EA 20 USD
76806-954 76806-956 76806-958
Geranyl formate 85%
Geranyl formate

Precaución: for Research Use Only

Formula: C₁₁H₁₈O₂
MDL Number: MFCD00021047
CAS Number: 105-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR