H-Asp-OH.Mg 98%

Supplier: Ambeed
Warning

A725422-1000G A725422-100G A725422-500G A725422-25G
76806-948EA 70.02 USD
76806-948 76806-950 76806-952 77308-214
H-Asp-OH.Mg 98%
H-Asp-OH.Mg

Precaución: for Research Use Only

Formula: C₈H₁₂MgN₂O₈
MW: 288.49 g/mol
MDL Number: MFCD00044935
CAS Number: 2068-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR