S-(Acetamidomethyl)-N-(tert-butoxycarbonyl)-L-cysteine 98%

Supplier: Ambeed
Warning

Synonyms: Boc-Cys(Acm)-OH

A725037-1G A725037-25G A725037-5G A725037-100G
76806-936EA 6.37 USD
76806-936 76806-938 76806-940 77669-952
S-(Acetamidomethyl)-N-(tert-butoxycarbonyl)-L-cysteine 98%
S-(Acetamidomethyl)-N-(tert-butoxycarbonyl)-L-cysteine

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₅S
MW: 292.35 g/mol
MDL Number: MFCD00038252
CAS Number: 19746-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR