4-tert-Butylbenzylamine 98%

Supplier: Ambeed
Warning

Synonyms: p-tert-Butylbenzylamine

A724509-100G A724509-5G
76806-934EA 115 USD
76806-934 77552-244
4-tert-Butylbenzylamine 98%
4-tert-Butylbenzylamine

Precaución: for Research Use Only

Formula: C₁₁H₁₇N
MW: 163.26 g/mol
MDL Number: MFCD00040754
CAS Number: 39895-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR