Isobutyrophenone 98%

Supplier: Ambeed
Warning

Synonyms: Isopropyl phenyl ketone, 2-Methyl-1-phenyl-1-propanone, Phenyl isopropyl ketone, α-Methylpropiophenone

A723571-100G A723571-10G A723571-1G A723571-25G A723571-5G
76806-918EA 82.76 USD
76806-918 76806-920 76806-922 76806-924 76806-926
Isobutyrophenone 98%
Isobutyrophenone

Precaución: for Research Use Only

Formula: (C₆H₅)C(O)CH(CH₃)₂
MW: 148.2 g/mol
MDL Number: MFCD00008917
CAS Number: 611-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR