L(+)-Diisopropyl tartrate 98%

Supplier: Ambeed
Warning

A723406-100G A723406-25G A723406-500G A723406-5G A723406-1000G
76806-912EA 36 USD
76806-912 76806-914 76806-916 77602-860 77669-950
L(+)-Diisopropyl tartrate 98%
L(+)-Diisopropyl tartrate

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₆
MDL Number: MFCD00064450
CAS Number: 2217-15-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR