Diethyl chloromalonate 96%

Supplier: Ambeed
Danger

Synonyms: Diethyl 2-chloromalonate

A723261-100G A723261-10G A723261-1G A723261-25G A723261-5G
76806-902EA 147 USD
76806-902 76806-904 76806-906 76806-908 76806-910
Diethyl chloromalonate 96%
Diethyl chloromalonate

Precaución: for Research Use Only

Formula: C₇H₁₁ClO₄
MDL Number: MFCD00009140
CAS Number: 14064-10-9
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR