di-Sodium succinate hexahydrate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Sodium succinate hexahydrate, Succinic acid disodium hexahydrate, 1,4-Butanedioic acid disodium salt hexahydrate, Sodium Succinate, Hexahydrate, Sodium succinate dibasic hexahydrate, Disodium succinate hexahydrate, Succinic acid disodium salt hexahydrate
A722854-1000G
A722854-100G
A722854-25G
A722854-500G
76806-894EA
18
USD
76806-894
76806-896
76806-898
76806-900
di-Sodium succinate hexahydrate 98%
di-Sodium succinate hexahydrate
Precaución: for Research Use Only
Formula:
NaOOCCH₂CH₂COONa·6H₂O MW: 270.14 g/mol Melting Pt: 300 |
MDL Number:
MFCD00149117 CAS Number: 6106-21-4 Merck Index: 13,08754 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...