Calcium ascorbate dihydrate 95%

Supplier: Ambeed
Warning

Synonyms: Calcium (R)-5-((S)-1,2-dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one salt dihydrate, Vitamin C calcium salt, L-Ascorbic acid calcium salt dihydrate, Calcium L-ascorbate dihydrate

A722666-1000G A722666-100G A722666-25G A722666-500G
76806-886EA 40.32 USD
76806-886 76806-888 76806-890 76806-892
Calcium ascorbate dihydrate 95%
Calcium ascorbate dihydrate

Precaución: for Research Use Only

Formula: C₁₂H₁₄CaO₁₂·2H₂O
MW: 426.34 g/mol
MDL Number: MFCD00149216
CAS Number: 5743-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR