1,1,3,3-Tetraphenyl-1,3-divinyldisiloxane 95% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A722621-1G A722621-250MG A722621-5G
76806-880EA 33 USD
76806-880 76806-882 76806-884
1,1,3,3-Tetraphenyl-1,3-divinyldisiloxane 95% stabilized
1,1,3,3-Tetraphenyl-1,3-divinyldisiloxane

Precaución: for Research Use Only

Formula: C₂₈H₂₆OSi₂
MDL Number: MFCD00271001
CAS Number: 18769-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR