Methyl-3-hydroxyhexanoate 95%

Supplier: Ambeed
Danger

Synonyms: Methyl 3-hydroxyhexanoate

A721450-5G A721450-1G
76806-866EA 25 USD
76806-866 77880-482
Methyl-3-hydroxyhexanoate 95%
Methyl 3-hydroxyhexanoate

Precaución: for Research Use Only

Formula: C₇H₁₄O₃
MDL Number: MFCD00083583
CAS Number: 21188-58-9
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR