Ethyl-3-cyano-2-hydroxy-6-methylisonicotinate 98%

Supplier: Ambeed
Warning

A721350-10G A721350-1G A721350-250MG A721350-5G
76806-858EA 227.99 USD
76806-858 76806-860 76806-862 76806-864
Ethyl-3-cyano-2-hydroxy-6-methylisonicotinate 98%
Ethyl-3-cyano-2-hydroxy-6-methylisonicotinate

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₃
MDL Number: MFCD00052274
CAS Number: 18619-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR