Hexadecyl acrylate 98% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A720933-100G A720933-25G A720933-500G
76806-844EA 91 USD
76806-844 76806-846 76806-848
Hexadecyl acrylate 98% stabilized
Hexadecyl acrylate

Precaución: for Research Use Only

Formula: C₁₉H₃₆O₂
MDL Number: MFCD00078441
CAS Number: 13402-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR