Ethyl-α-chlorophenylacetate 97%

Supplier: Ambeed
Danger

Synonyms: Ethyl 2-chloro-2-phenylacetate

A717100-10G A717100-1G A717100-25G A717100-5G
76806-816EA 73.21 USD
76806-816 76806-818 76806-820 76806-822
Ethyl-α-chlorophenylacetate 97%
Ethyl α-chlorophenylacetate

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₂
MW: 198.6461 g/mol
MDL Number: MFCD00013675
CAS Number: 4773-33-5
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR