Diindium triselenide 99.99% ~60M

Supplier: Ambeed
Danger

A716555-1G A716555-25G A716555-5G
76806-810EA 18 USD
76806-810 76806-812 76806-814
Diindium triselenide 99.99% ~60M
Diindium triselenide

Precaución: for Research Use Only

Formula: In₂Se₃
MDL Number: MFCD00016147
CAS Number: 12056-07-4
UN: 3288
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR