(3-(tert-Butylcarbamoyl)-5-nitrophenyl)boronic acid 96%
Supplier: Ambeed
|
Warning
|
A716105-1G
A716105-5G
76806-806EA
61.54
USD
76806-806
76806-808
(3-(tert-Butylcarbamoyl)-5-nitrophenyl)boronic acid 96%
(3-(tert-Butylcarbamoyl)-5-nitrophenyl)boronic acid
Precaución: for Research Use Only
Formula:
C₁₁H₁₅BN₂O₅ MW: 266.06 g/mol |
MDL Number:
MFCD07783879 CAS Number: 871332-87-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...