Dihexylamine 98%

Supplier: Ambeed
Danger

A714544-25G A714544-100G A714544-500G
76806-798EA 9.55 USD
76806-798 77595-116 77627-460
Dihexylamine 98%
Dihexylamine

Precaución: for Research Use Only

Formula: C₁₂H₂₇N
MW: 185.35 g/mol
MDL Number: MFCD00009521
CAS Number: 143-16-8
UN: 2922
ADR: 8,6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR