2-Methylleucine 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Amino-2,4-dimethylpentanoic acid, H-α-Me-Leu-OH, (S)-alpha-Methylleucine

A712606-100MG A712606-250MG
76806-794EA 101.85 USD
76806-794 76806-796
2-Methylleucine 97%
2-Methylleucine

Precaución: for Research Use Only

Formula: C₇H₁₅NO₂
MW: 145.202 g/mol
MDL Number: MFCD01716279
CAS Number: 105743-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR