Methyl 2-(1-(4-chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl)acetate 98%
Supplier: Ambeed
|
Warning
|
A706743-100MG
A706743-1G
A706743-250MG
A706743-5G
76806-754EA
59.42
USD
76806-754
76806-756
76806-758
76806-760
Methyl 2-(1-(4-chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl)acetate 98%
Methyl 2-(1-(4-chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl)acetate
Precaución: for Research Use Only
Formula:
C₂₀H₁₈ClNO₄ MW: 371.81 g/mol |
MDL Number:
MFCD01719291 CAS Number: 1601-18-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...