Ethyl pyrazolo[1,5-a]pyridine-3-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethylpyrazolo[1,5-a]pyridine-3-carboxylate

A706319-1G A706319-250MG A706319-5G
76806-744EA 36.08 USD
76806-744 76806-746 77669-906
Ethyl pyrazolo[1,5-a]pyridine-3-carboxylate 98%
Ethyl pyrazolo[1,5-a]pyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MW: 190.1986 g/mol
MDL Number: MFCD06411542
CAS Number: 16205-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR