Fmoc-D-Bpa-OH 98%

Supplier: Ambeed
Warning

A705099-1G A705099-250MG A705099-5G
76806-738EA 26 USD
76806-738 76806-740 76806-742
Fmoc-D-Bpa-OH 98%
Fmoc-D-Bpa-OH

Precaución: for Research Use Only

Formula: C₃₁H₂₅NO₅
MDL Number: MFCD00237666
CAS Number: 117666-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR