Fmoc-(3-aminomethylphenyl)acetic acid 95%

Supplier: Ambeed
Warning

A705016-1G A705016-100MG A705016-250MG
76806-736EA 150 USD
76806-736 77880-461 77880-462
Fmoc-(3-aminomethylphenyl)acetic acid 95%
Fmoc-(3-aminomethylphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₂₄H₂₁NO₄
MDL Number: MFCD06656460
CAS Number: 631915-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR