Methyl cis-13-docosenoate 90%

Supplier: Ambeed
Danger

A701664-1G A701664-25G A701664-5G
76806-724EA 7 USD
76806-724 76806-726 76806-728
Methyl cis-13-docosenoate 90%
Methyl cis-13-docosenoate

Precaución: for Research Use Only

Formula: C₂₃H₄₄O₂
MDL Number: MFCD00027343
CAS Number: 1120-34-9
UN: 3272
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR