3-(Trifluoromethoxy)cinnamic acid 98%

Supplier: Ambeed
Danger

Synonyms: 3-(3-(Trifluoromethoxy)phenyl)acrylic acid

A700307-1G A700307-25G A700307-5G
76806-710EA 12.74 USD
76806-710 76806-712 76806-714
3-(Trifluoromethoxy)cinnamic acid 98%
3-(Trifluoromethoxy)cinnamic acid

Precaución: for Research Use Only

Formula: C₁₀H₇F₃O₃
MW: 232.16 g/mol
MDL Number: MFCD00066337
CAS Number: 168833-80-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR