Ethyl-2,4,6-triisopropylbenzoate 97%

Supplier: Ambeed
Warning

A698399-1G A698399-250MG A698399-5G
76806-702EA 62 USD
76806-702 76806-704 76806-706
Ethyl-2,4,6-triisopropylbenzoate 97%
Ethyl-2,4,6-triisopropylbenzoate

Precaución: for Research Use Only

Formula: C₁₈H₂₈O₂
MDL Number: MFCD03788745
CAS Number: 63846-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR