2-Amino-3-(4-(tert-butyl)phenyl)propanoic acid 95%

Supplier: Ambeed
Warning

A695179-100MG A695179-1G A695179-250MG A695179-5G
76806-694EA 21.22 USD
76806-694 76806-696 76806-698 76806-700
2-Amino-3-(4-(tert-butyl)phenyl)propanoic acid 95%
2-Amino-3-(4-(tert-butyl)phenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO₂
MW: 221.29 g/mol
MDL Number: MFCD05227971
CAS Number: 98708-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR