EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
EGTA, Ethylenebis(oxyethylenenitrilo)tetra(acetic acid), Kestranal® FE PA, Idranal® VI, Ethylene glycol-O,O'-bis(2-aminoethyl)-N,N,N',N'-tetraacetic acid, Ethylenebis(oxyethylenenitrilo)tetraacetic acid, Ethylene Glycol Bis(2-aminoethyl Ether)-N,N,N,N-tetraacetic acid, 3,12-Bis(carboxymethyl)-6,9-dioxa-3,12-diazatetradecanedioic acid, Titriplex® VI
A694728-100G
A694728-1MG
A694728-25G
A694728-5G
A694728-500G
76806-686EA
67
USD
76806-686
76806-688
76806-690
76806-692
77627-404
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) 95%
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid)
Precaución: for Research Use Only
Formula:
C₁₄H₂₄N₂O₁₀ MW: 380.35 g/mol |
MDL Number:
MFCD00004291 CAS Number: 67-42-5 Merck Index: 13,03560 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...