(+)-Dimethyl-L-tartrate 97%

Supplier: Ambeed
Warning

Synonyms: Dimethyl (2R,3R)-2,3-dihydroxysuccinate, (R,R)-(+)-Tartaric acid dimethyl ester, (R,R)-(+)-Dimethyl tartrate, L(+)-Tartaric acid dimethyl ester, (+)-Dimethyl L-tartrate

A694721-100G A694721-25G A694721-500G A694721-5G
76806-670EA 46 USD
76806-670 76806-672 76806-674 76806-676
(+)-Dimethyl-L-tartrate 97%
(+)-Dimethyl-L-tartrate

Precaución: for Research Use Only

Formula: C₆H₁₀O₆
MW: 178.14 g/mol
Boiling Pt: 245
MDL Number: MFCD00064437
CAS Number: 608-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR