3,5-Bis(trifluoromethyl)cinnamic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-(3,5-Bis(trifluoromethyl)phenyl)acrylic acid

A694653-100G A694653-5G
76806-666EA 107.16 USD
76806-666 76806-668
3,5-Bis(trifluoromethyl)cinnamic acid 95%
3,5-Bis(trifluoromethyl)cinnamic acid

Precaución: for Research Use Only

Formula: C₁₁H₆F₆O₂
MW: 284.1546 g/mol
MDL Number: MFCD00799487
CAS Number: 155814-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR